Assay Details
Target Gene Details
Entrez Gene ID: | 389 |
Gene Name: | ras homolog family member C |
Gene Aliases: |
ARH9, ARHC, H9, RHOH9 |
Location: |
Chr.1:112701127-112707403 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOC | NM_001042678.1 | NP_001036143.1 | ||
| NM_001042679.1 | NP_001036144.1 | |||
| NM_175744.4 | NP_786886.1 | |||
| AK094474.1 | ||||
| BC007245.1 | AAH07245.1 | |||
| BC009177.2 | AAH09177.1 | |||
| BQ231012.1 | ||||
| BX648784.1 | ||||
| CN289691.1 | ||||
| CX872299.1 | ||||
| L25081.1 | AAC33179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998794 | Chr.1:112704008 - 112767796 on Build GRCh38 | Gain |
|
| esv27598 | Chr.1:112703696 - 112808153 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map