Assay Details
Target Gene Details
Entrez Gene ID: | 403314 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4 |
Gene Aliases: |
C1orf169 |
Location: |
Chr.1:183646275-183653313 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC4 | NM_203454.2 | 2 | 358 | NP_982279.1 |
| AK098557.1 | 2 | 366 | BAC05332.1 | |
| BC021711.2 | 2 | 358 | AAH21711.1 |
Target Gene Details
Entrez Gene ID: | 23179 |
Gene Name: | ral guanine nucleotide dissociation stimulator like 1 |
Gene Aliases: |
RGL |
Location: |
Chr.1:183636047-183928551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGL1 | NM_001297669.1 | NP_001284598.1 | ||
| NM_001297670.1 | NP_001284599.1 | |||
| NM_015149.4 | NP_055964.3 | |||
| XM_011509339.2 | XP_011507641.1 | |||
| XM_011509341.1 | XP_011507643.1 | |||
| XM_011509342.2 | XP_011507644.1 | |||
| AF186780.1 | AAF67281.1 | |||
| AK295179.1 | ||||
| AK295611.1 | ||||
| AK316526.1 | ||||
| BC136591.1 | ||||
| BP235344.1 | ||||
| BX537416.1 | CAD97658.1 | |||
| DC333918.1 |
More Information
Set Membership: |
|

Genomic Map