Assay Details
Target Gene Details
Entrez Gene ID: | 54957 |
Gene Name: | thioredoxin like 4B |
Gene Aliases: |
DLP, Dim2 |
Location: |
Chr.16:72044261-72094316 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNL4B | NM_001142317.1 | 4 | 697 | NP_001135789.1 |
| NM_001142318.1 | 4 | 595 | NP_001135790.1 | |
| NM_001324354.1 | NP_001311283.1 | |||
| NM_001324355.1 | 5 | 846 | NP_001311284.1 | |
| NM_017853.2 | 4 | 746 | NP_060323.1 | |
| XM_017023377.1 | XP_016878866.1 | |||
| AK000518.1 | 4 | 721 | BAA91224.1 | |
| AK222889.1 | 4 | 713 | BAD96609.1 | |
| AK308891.1 | ||||
| AY566808.1 | AAS68520.1 | |||
| BC009646.1 | 4 | 687 | AAH09646.1 | |
| CD517462.1 | 4 | 643 | ||
| CR457240.1 | CAG33521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428329 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
| dgv3017n100 | Chr.16:72040933 - 72757730 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map