Assay Details
Target Gene Details
Entrez Gene ID: | 84527 |
Gene Name: | zinc finger protein 559 |
Gene Aliases: |
NBLA00121 |
Location: |
Chr.19:9323772-9343845 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF559 | NM_001202406.1 | NP_001189335.1 | ||
NM_001202407.1 | NP_001189336.1 | |||
NM_001202408.1 | NP_001189337.1 | |||
NM_001202409.1 | NP_001189338.1 | |||
NM_001202410.1 | NP_001189339.1 | |||
NM_001202411.1 | NP_001189340.1 | |||
NM_001202412.1 | NP_001189341.1 | |||
NM_032497.2 | NP_115886.1 | |||
AB075488.1 | 1 | 76 | BAE45747.1 | |
AK027700.1 | ||||
AK056489.1 | ||||
AK092348.1 | ||||
AK298169.1 | ||||
AK308517.1 | ||||
AK309952.1 | ||||
AK310020.1 | ||||
AK310117.1 | ||||
AK310781.1 | ||||
BC006436.1 | AAH06436.1 | |||
BP337457.1 | ||||
BP417021.1 | ||||
CF597362.1 | ||||
DA509657.1 | ||||
DA743493.1 | ||||
DA774704.1 | ||||
DB158737.1 | ||||
DB288007.1 |
Target Gene Details
Entrez Gene ID: | 100529215 |
Gene Name: | ZNF559-ZNF177 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:9324226-9382617 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF559-ZNF177 | NM_001172650.2 | NP_001166121.1 | ||
NM_001202425.1 | NP_001189354.1 | |||
U37251.1 | AAB09748.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
