Assay Details
Target Gene Details
Entrez Gene ID: | 165545 |
Gene Name: | DEAQ-box RNA dependent ATPase 1 |
Gene Aliases: |
- |
Location: |
Chr.2:74518131-74526281 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DQX1 | NM_133637.2 | NP_598376.2 | ||
| XM_011532644.1 | XP_011530946.1 | |||
| XM_011532646.2 | XP_011530948.1 | |||
| XM_017003520.1 | XP_016859009.1 | |||
| AK074337.1 | BAB85056.1 | |||
| AK092420.1 | ||||
| BC075018.2 | AAH75018.1 | |||
| DA852016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map