Assay Details
Target Gene Details
Entrez Gene ID: | 139378 |
Gene Name: | adhesion G protein-coupled receptor G4 |
Gene Aliases: |
GPR112, PGR17, RP1-299I16 |
Location: |
Chr.X:136300963-136416888 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG4 | NM_153834.3 | 6 | 2311 | NP_722576.3 |
| XM_011531269.2 | 5 | 2317 | XP_011529571.1 | |
| XM_011531271.2 | 3 | 2049 | XP_011529573.1 | |
| AY140954.1 | 2 | 1828 | AAN46668.1 | |
| AY255581.1 | 1 | 994 | AAO85093.1 | |
| AY882585.1 | 6 | 2311 | AAW78659.1 | |
| CR936801.1 | 3 | 1555 |
More Information
Set Membership: |
|

Genomic Map