Assay Details
Target Gene Details
Entrez Gene ID: | 247 |
Gene Name: | arachidonate 15-lipoxygenase, type B |
Gene Aliases: |
15-LOX-2 |
Location: |
Chr.17:8039040-8049134 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX15B | NM_001039130.1 | 13 | 2353 | NP_001034219.1 |
| NM_001039131.1 | 12 | 2218 | NP_001034220.1 | |
| NM_001141.2 | 14 | 2440 | NP_001132.2 | |
| AK298095.1 | ||||
| BC035217.1 | 14 | 2382 | AAH35217.1 | |
| BC063647.1 | 14 | 2396 | AAH63647.1 | |
| CA307212.1 | 1 | 271 | ||
| U78294.1 | 14 | 2395 | AAB61706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528325 | Chr.17:8048501 - 8051047 on Build GRCh38 | Loss |
|
| nsv833352 | Chr.17:8029719 - 8187575 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map