Assay Details
Target Gene Details
Entrez Gene ID: | 55601 |
Gene Name: | DEXD/H-box helicase 60 |
Gene Aliases: |
- |
Location: |
Chr.4:168216291-168318807 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX60 | NM_017631.5 | 26 | 3779 | NP_060101.3 |
| XM_011532103.2 | 26 | 3779 | XP_011530405.1 | |
| XM_011532104.2 | XP_011530406.1 | |||
| AK001649.1 | BAA91809.1 | |||
| AK307404.1 | ||||
| BC038115.2 | 26 | 3724 | AAH38115.1 | |
| BX648655.1 | 6 | 620 | CAE46197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4596 | Chr.4:168228336 - 168263735 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map