Assay Details
Target Gene Details
Entrez Gene ID: | 5641 |
Gene Name: | legumain |
Gene Aliases: |
AEP, LGMN1, PRSC1 |
Location: |
Chr.14:92703807-92748702 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGMN | NM_001008530.2 | NP_001008530.1 | ||
| NM_005606.6 | NP_005597.3 | |||
| XM_005267862.3 | XP_005267919.1 | |||
| XM_005267863.3 | XP_005267920.1 | |||
| XM_011536970.1 | XP_011535272.1 | |||
| XM_017021463.1 | XP_016876952.1 | |||
| XM_017021464.1 | XP_016876953.1 | |||
| XM_017021465.1 | XP_016876954.1 | |||
| XM_017021466.1 | XP_016876955.1 | |||
| XM_017021467.1 | XP_016876956.1 | |||
| AK225822.1 | ||||
| AK291534.1 | ||||
| AK291560.1 | ||||
| AK297074.1 | ||||
| AK297780.1 | ||||
| BC003061.2 | AAH03061.1 | |||
| BX161380.1 | CAD61872.1 | |||
| BX161422.1 | CAD61895.1 | |||
| BX161434.1 | CAD61906.1 | |||
| CR457406.1 | CAG33687.1 | |||
| D55696.1 | BAA09530.1 | |||
| Y09862.1 | CAA70989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv417e214 | Chr.14:92692074 - 92763860 on Build GRCh38 | Gain |
|
| nsv565525 | Chr.14:92704648 - 92752613 on Build GRCh38 | Loss |
|
| nsv456399 | Chr.14:92685875 - 92724438 on Build GRCh38 | Loss |
|
| nsv1160201 | Chr.14:92634527 - 92716121 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map