Assay Details
Target Gene Details
Entrez Gene ID: | 2903 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2A |
Gene Aliases: |
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A |
Location: |
Chr.16:9753404-10183364 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2A | NM_000833.4 | 14 | 4854 | NP_000824.1 |
| NM_001134407.2 | 13 | 5092 | NP_001127879.1 | |
| NM_001134408.2 | 14 | 4410 | NP_001127880.1 | |
| XM_011522458.2 | 12 | 4340 | XP_011520760.1 | |
| XM_011522461.2 | 15 | 5121 | XP_011520763.1 | |
| XM_017023172.1 | 12 | 4965 | XP_016878661.1 | |
| XM_017023173.1 | 13 | 4622 | XP_016878662.1 | |
| AK295620.1 | 13 | 3842 | ||
| U09002.1 | 13 | 4699 | AAB60343.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75272984] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map