Assay Details
Target Gene Details
Entrez Gene ID: | 252839 |
Gene Name: | transmembrane protein 9 |
Gene Aliases: |
DERM4, TMEM9A |
Location: |
Chr.1:201134771-201171625 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM9 | NM_001288564.1 | 6 | 858 | NP_001275493.1 |
| NM_001288565.1 | 5 | 1361 | NP_001275494.1 | |
| NM_001288566.1 | 6 | 959 | NP_001275495.1 | |
| NM_001288567.1 | 6 | 1173 | NP_001275496.1 | |
| NM_001288568.1 | 6 | 1011 | NP_001275497.1 | |
| NM_001288569.1 | 6 | 872 | NP_001275498.1 | |
| NM_001288570.1 | 6 | 885 | NP_001275499.1 | |
| NM_001288571.1 | 6 | 951 | NP_001275500.1 | |
| NM_016456.4 | 6 | 876 | NP_057540.1 | |
| NR_109920.1 | 7 | 1103 | ||
| NR_109921.1 | 5 | 1330 | ||
| XM_011509385.1 | 6 | 935 | XP_011507687.1 | |
| XM_011509386.2 | 6 | 910 | XP_011507688.1 | |
| XM_011509387.2 | 6 | 798 | XP_011507689.1 | |
| AF151020.1 | 7 | 848 | AAF36106.1 | |
| AK054883.1 | 6 | 831 | BAB70820.1 | |
| AK075335.1 | 6 | 786 | BAC11554.1 | |
| AK091726.1 | 5 | 1246 | ||
| AK094588.1 | 2 | 2179 | ||
| AK303843.1 | 6 | 897 | ||
| AL136600.1 | 6 | 815 | CAB66535.1 | |
| AY359012.1 | 6 | 850 | AAQ89371.1 | |
| BC001106.2 | 6 | 800 | AAH01106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428301 | Chr.1:201059216 - 201191346 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map