Assay Details
Target Gene Details
Entrez Gene ID: | 54880 |
Gene Name: | BCL6 corepressor |
Gene Aliases: |
ANOP2, MAA2, MCOPS2 |
Location: |
Chr.X:40051246-40177390 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCOR | NM_001123383.1 | 14 | 5158 | NP_001116855.1 |
| NM_001123384.1 | 13 | 5104 | NP_001116856.1 | |
| NM_001123385.1 | 14 | 5189 | NP_001116857.1 | |
| NM_017745.5 | 14 | 5087 | NP_060215.4 | |
| XM_005272616.1 | 14 | 5321 | XP_005272673.1 | |
| XM_005272618.3 | 14 | 4987 | XP_005272675.1 | |
| XM_005272619.3 | 13 | 5593 | XP_005272676.1 | |
| XM_005272620.3 | 13 | 5491 | XP_005272677.1 | |
| XM_006724536.3 | 14 | 6423 | XP_006724599.1 | |
| XM_011543929.2 | 14 | 5097 | XP_011542231.1 | |
| XM_011543930.1 | 14 | 5110 | XP_011542232.1 | |
| XM_011543931.2 | 14 | 5220 | XP_011542233.1 | |
| XM_017029615.1 | 14 | 6321 | XP_016885104.1 | |
| XM_017029616.1 | 14 | 5647 | XP_016885105.1 | |
| AF317391.1 | 14 | 5046 | AAG41429.1 | |
| AK000292.1 | 6 | 916 | BAA91061.1 | |
| AK074286.1 | 10 | 4095 | BAB85037.1 | |
| AK095360.1 | 9 | 1798 | ||
| AK225581.1 | 6 | 917 | ||
| AY316592.1 | 14 | 5148 | AAR08265.1 | |
| BC009675.2 | 5 | 715 | AAH09675.2 | |
| BC063536.1 | 3 | 271 | AAH63536.1 | |
| BC114220.1 | 13 | 5104 | AAI14221.1 |
More Information
Set Membership: |
|

Genomic Map