Assay Details
Target Gene Details
Entrez Gene ID: | 665 |
Gene Name: | BCL2 interacting protein 3 like |
Gene Aliases: |
BNIP3a, NIX |
Location: |
Chr.8:26383007-26413128 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIP3L | NM_004331.2 | 6 | 1551 | NP_004322.1 |
| XM_005273617.3 | 6 | 1455 | XP_005273674.1 | |
| AF536326.1 | AAN04051.1 | |||
| AL132665.1 | 6 | 1525 | CAI46217.1 | |
| CD244492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831273 | Chr.8:26358989 - 26513275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map