Assay Details
Target Gene Details
Entrez Gene ID: | 23759 |
Gene Name: | peptidylprolyl isomerase like 2 |
Gene Aliases: |
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60 |
Location: |
Chr.22:21665984-21698276 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL2 | NM_001317996.1 | 21 | 3474 | NP_001304925.1 |
| NM_014337.3 | 20 | 3844 | NP_055152.1 | |
| NM_148175.2 | 21 | 2594 | NP_680480.1 | |
| NM_148176.2 | 21 | 2452 | NP_680481.1 | |
| XM_005261448.3 | 21 | 2456 | XP_005261505.1 | |
| XM_011530041.2 | 21 | 3470 | XP_011528343.1 | |
| XM_011530042.2 | 21 | 2590 | XP_011528344.1 | |
| XM_011530043.2 | 21 | 2448 | XP_011528345.1 | |
| XM_011530044.2 | 21 | 2586 | XP_011528346.1 | |
| XM_011530046.2 | 21 | 2452 | XP_011528348.1 | |
| XM_011530047.2 | 21 | 2590 | XP_011528349.1 | |
| XM_011530049.2 | 22 | 3520 | XP_011528351.1 | |
| XM_011530050.2 | 20 | 3383 | XP_011528352.1 | |
| XM_011530051.2 | 21 | 3410 | XP_011528353.1 | |
| XM_017028705.1 | 12 | 2716 | XP_016884194.1 | |
| XM_017028706.1 | 12 | 1702 | XP_016884195.1 | |
| AA743164.1 | 1 | 285 | ||
| AK097249.1 | 3 | 1488 | ||
| AK127259.1 | 22 | 3956 | ||
| BC000022.2 | 21 | 2414 | AAH00022.1 | |
| BC028385.2 | 21 | 3254 | AAH28385.1 | |
| BM981665.1 | ||||
| U37220.1 | 12 | 1680 | AAC50377.1 | |
| U37221.1 | 12 | 1853 | AAC50378.1 |
Target Gene Details
Entrez Gene ID: | 29799 |
Gene Name: | yippee like 1 |
Gene Aliases: |
FKSG3 |
Location: |
Chr.22:21697536-21735834 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YPEL1 | NM_013313.4 | 5 | 4218 | NP_037445.1 |
| NR_130910.1 | 5 | 4602 | ||
| AB098734.1 | 5 | 4218 | BAD51375.1 | |
| AK096530.1 | 1 | 2080 | ||
| BC034486.1 | 5 | 4076 | AAH34486.1 | |
| CR933630.1 | 4 | 8321 | CAI45937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1293e212 | Chr.22:21685588 - 21699487 on Build GRCh38 | Loss |
|
| nsv510804 | Chr.22:21670242 - 21778475 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map