Assay Details
Target Gene Details
Entrez Gene ID: | 8790 |
Gene Name: | fucose-1-phosphate guanylyltransferase |
Gene Aliases: |
GFPP |
Location: |
Chr.1:74198212-74208702 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FPGT | NM_001199328.2 | 5 | 1773 | NP_001186257.2 |
| NM_001199329.2 | 4 | 1718 | NP_001186258.2 | |
| NM_003838.4 | 4 | 2535 | NP_003829.3 | |
| AF017445.1 | 4 | 2505 | AAC73005.1 | |
| AK294948.1 | ||||
| BC032308.1 | 4 | 2491 | AAH32308.1 |
Target Gene Details
Entrez Gene ID: | 100526835 |
Gene Name: | FPGT-TNNI3K readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:74198212-74544432 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FPGT-TNNI3K | NM_001112808.2 | NP_001106279.2 | ||
| NM_001199327.1 | NP_001186256.2 | |||
| AB208941.1 | BAD92178.1 | |||
| BC032865.1 | AAH32865.1 | |||
| BX640903.1 | CAE45949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv28e214 | Chr.1:74183056 - 74208995 on Build GRCh38 | Gain |
|
| esv3893723 | Chr.1:73643288 - 74246497 on Build GRCh38 | Gain |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|
| nsv522147 | Chr.1:73872847 - 74518151 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map