Assay Details
Target Gene Details
Entrez Gene ID: | 121643 |
Gene Name: | forkhead box N4 |
Gene Aliases: |
- |
Location: |
Chr.12:109277978-109309406 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXN4 | NM_213596.2 | 10 | 1910 | NP_998761.2 |
| XM_011537922.2 | 10 | 2099 | XP_011536224.1 | |
| XM_017018818.1 | 8 | 2885 | XP_016874307.1 | |
| AK093286.1 | ||||
| AK131519.1 | 6 | 1517 | BAD18661.1 | |
| BC113073.1 | 3 | 910 | ||
| BC146825.1 | 10 | 1911 |
More Information
Set Membership: |
|

Genomic Map