Assay Details
Target Gene Details
Entrez Gene ID: | 2888 |
Gene Name: | growth factor receptor bound protein 14 |
Gene Aliases: |
- |
Location: |
Chr.2:164492406-164621850 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRB14 | NM_001303422.1 | 9 | 1080 | NP_001290351.1 |
| NM_004490.2 | 10 | 1709 | NP_004481.2 | |
| XM_017003899.1 | 10 | 1146 | XP_016859388.1 | |
| AK301961.1 | 9 | 1025 | ||
| AK302129.1 | ||||
| AK307744.1 | 10 | 1222 | ||
| AK313153.1 | 10 | 1338 | ||
| BC053559.1 | 10 | 1179 | AAH53559.1 | |
| L76687.1 | 10 | 1708 | AAC15861.1 |
More Information
Set Membership: |
|

Genomic Map