Assay Details
Target Gene Details
Entrez Gene ID: | 57595 |
Gene Name: | PDZ domain containing 4 |
Gene Aliases: |
LNX5, LU1, PDZK4, PDZRN4L |
Location: |
Chr.X:153802166-153830567 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZD4 | NM_001303512.1 | 8 | 3112 | NP_001290441.1 |
| NM_001303513.1 | 9 | 3355 | NP_001290442.1 | |
| NM_001303514.1 | 6 | 2767 | NP_001290443.1 | |
| NM_001303515.1 | 8 | 2947 | NP_001290444.1 | |
| NM_001303516.1 | 8 | 2929 | NP_001290445.1 | |
| NM_032512.3 | 8 | 3094 | NP_115901.2 | |
| AB103478.1 | 8 | 3075 | BAD16631.1 | |
| AK091444.1 | 8 | 2820 | BAC03666.1 | |
| AK123229.1 | ||||
| AK123822.1 | 8 | 2947 | ||
| AK127016.1 | 8 | 3094 | ||
| BC002606.2 | 1 | 1225 | AAH02606.2 | |
| BC080584.1 | 1 | 1225 | AAH80584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|

Genomic Map