Assay Details
Target Gene Details
Entrez Gene ID: | 55929 |
Gene Name: | DNA methyltransferase 1 associated protein 1 |
Gene Aliases: |
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4 |
Location: |
Chr.1:44213428-44220679 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMAP1 | NM_001034023.1 | NP_001029195.1 | ||
| NM_001034024.1 | NP_001029196.1 | |||
| NM_019100.4 | NP_061973.1 | |||
| XM_017001808.1 | XP_016857297.1 | |||
| AB037846.1 | ||||
| AK021605.1 | BAB13854.1 | |||
| AK054954.1 | ||||
| AK092898.1 | ||||
| AK096252.1 | ||||
| AK289366.1 | ||||
| AK298793.1 | ||||
| AL136657.1 | CAB66592.1 | |||
| AL137200.1 | CAB69910.1 | |||
| BC002855.2 | AAH02855.1 | |||
| BC008053.2 | AAH08053.1 | |||
| BU616029.1 | ||||
| BX537895.1 | CAD97886.1 |
More Information
Set Membership: |
|

Genomic Map