Assay Details
Target Gene Details
Entrez Gene ID: | 84824 |
Gene Name: | Fc receptor like A |
Gene Aliases: |
FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB |
Location: |
Chr.1:161706972-161714352 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRLA | NM_001184866.1 | 6 | 1460 | NP_001171795.1 |
| NM_001184867.1 | 4 | 1175 | NP_001171796.1 | |
| NM_001184870.1 | 4 | 1040 | NP_001171799.1 | |
| NM_001184871.1 | 2 | 737 | NP_001171800.1 | |
| NM_001184872.1 | 3 | 1022 | NP_001171801.1 | |
| NM_001184873.1 | 3 | 890 | NP_001171802.1 | |
| NM_032738.3 | 5 | 1442 | NP_116127.3 | |
| XM_006711581.3 | 5 | 1143 | XP_006711644.1 | |
| XM_011510064.1 | 6 | 1200 | XP_011508366.1 | |
| XM_011510065.1 | 5 | 948 | XP_011508367.1 | |
| XM_011510066.1 | 4 | 1072 | XP_011508368.1 | |
| AF329489.1 | 5 | 1165 | AAL23899.1 | |
| AF329491.1 | 3 | 785 | AAL23901.1 | |
| AF329493.1 | AAL23903.1 | |||
| AF329494.1 | AAL23904.1 | |||
| AF329495.1 | AAL23905.1 | |||
| AF390036.1 | 3 | 567 | AAL27307.1 | |
| AF426461.1 | 5 | 1210 | AAL58111.1 | |
| AF531423.1 | 5 | 1442 | AAM97591.1 | |
| AY091642.2 | 6 | 1167 | AAM13980.2 | |
| AY358348.1 | 5 | 1157 | AAQ88714.1 | |
| BC006521.2 | 5 | 1257 | ||
| BX112608.1 | 2 | 421 | ||
| BX649184.1 | 3 | 2000 | CAE46202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map