Assay Details
Target Gene Details
Entrez Gene ID: | 4325 |
Gene Name: | matrix metallopeptidase 16 |
Gene Aliases: |
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP |
Location: |
Chr.8:88037232-88327488 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP16 | NM_005941.4 | 10 | 3690 | NP_005932.2 |
| AL832595.1 | 11 | 3729 | ||
| U79292.1 | 1 | 1046 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617938 | Chr.8:88025337 - 88043428 on Build GRCh38 | Gain |
|
| esv2759625 | Chr.8:87951601 - 88121475 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map