Assay Details
Target Gene Details
Entrez Gene ID: | 5587 |
Gene Name: | protein kinase D1 |
Gene Aliases: |
PKC-MU, PKCM, PKD, PRKCM |
Location: |
Chr.14:29576479-29927693 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKD1 | NM_002742.2 | 18 | 2892 | NP_002733.2 |
| XM_005267859.1 | 19 | 2850 | XP_005267916.1 | |
| XM_011536964.2 | 19 | 5272 | XP_011535266.1 | |
| XM_011536965.2 | 19 | 7322 | XP_011535267.1 | |
| XM_017021461.1 | 19 | 2627 | XP_016876950.1 | |
| XM_017021462.1 | 19 | 2589 | XP_016876951.1 | |
| AK314170.1 | ||||
| BX645735.1 | 1 | 202 | ||
| X75756.1 | 18 | 2946 | CAA53384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526536 | Chr.14:28322483 - 30669172 on Build GRCh38 | Gain |
|
| nsv564160 | Chr.14:29030900 - 29631166 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map