Assay Details
Target Gene Details
Entrez Gene ID: | 94160 |
Gene Name: | ATP binding cassette subfamily C member 12 |
Gene Aliases: |
MRP9 |
Location: |
Chr.16:48082973-48146770 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC12 | NM_033226.2 | 29 | 5104 | NP_150229.2 |
| AY040220.1 | 29 | 5104 | AAK76740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761906 | Chr.16:48075816 - 48091944 on Build GRCh38 | Loss |
|
| nsv520355 | Chr.16:47390917 - 48130866 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114144417] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map