Assay Details
Target Gene Details
Entrez Gene ID: | 57132 |
Gene Name: | charged multivesicular body protein 1B |
Gene Aliases: |
C10orf2, C18-ORF2, C18orf2, CHMP1.5, Vps46-2, Vps46B, hVps46-2 |
Location: |
Chr.18:11851390-11854449 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP1B | NM_020412.4 | 1 | 51 | NP_065145.2 |
| AF281064.1 | AAG01449.1 | |||
| AK314068.1 | 1 | 46 | ||
| BC012733.2 | ||||
| BC065933.1 | AAH65933.1 | |||
| BX494191.1 | 1 | 52 |
Target Gene Details
Entrez Gene ID: | 2774 |
Gene Name: | G protein subunit alpha L |
Gene Aliases: |
DYT25 |
Location: |
Chr.18:11688956-11885685 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAL | NM_001142339.2 | NP_001135811.1 | ||
| NM_001261443.1 | NP_001248372.1 | |||
| NM_182978.3 | NP_892023.1 | |||
| AF493893.1 | AAM12607.1 | |||
| AK090868.1 | BAC03535.1 | |||
| AK290054.1 | ||||
| AK295088.1 | ||||
| AK313902.1 | ||||
| BC050021.1 | AAH50021.1 | |||
| BP230966.1 | ||||
| L10665.1 | AAC37535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| esv3641815 | Chr.18:11840726 - 11854124 on Build GRCh38 | Gain |
|
| nsv833590 | Chr.18:11757330 - 11924540 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73409565] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map