Assay Details
Target Gene Details
Entrez Gene ID: | 57678 |
Gene Name: | glycerol-3-phosphate acyltransferase, mitochondrial |
Gene Aliases: |
GPAT, GPAT1 |
Location: |
Chr.10:112149864-112215395 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAM | NM_001244949.1 | NP_001231878.1 | ||
| NM_020918.5 | NP_065969.3 | |||
| XM_005269998.1 | XP_005270055.1 | |||
| AB046780.1 | BAB13386.1 | |||
| AK172782.1 | BAD18763.1 | |||
| AK172799.1 | BAD18772.1 | |||
| AL598680.1 | ||||
| AL832464.1 | 19 | 3487 | ||
| AL833061.2 | CAD89967.1 | |||
| AL833093.2 | CAD89932.1 | |||
| BC030783.1 | AAH30783.1 | |||
| BX647451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508608 | Chr.10:112030892 - 112158412 on Build GRCh38 | Deletion |
|
| nsv831989 | Chr.10:112112194 - 112301601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map