Assay Details
Target Gene Details
Entrez Gene ID: | 27429 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
OMI, PARK13, PRSS25 |
Location: |
Chr.2:74529405-74533556 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA2 | NM_001321727.1 | 7 | 2390 | NP_001308656.1 |
| NM_001321728.1 | 7 | 2360 | NP_001308657.1 | |
| NM_013247.4 | 8 | 2456 | NP_037379.1 | |
| NM_145074.2 | 6 | 2165 | NP_659540.1 | |
| NR_135769.1 | 8 | 2496 | ||
| NR_135770.1 | 9 | 1924 | ||
| NR_135771.1 | 9 | 1908 | ||
| NR_135772.1 | 9 | 1928 | ||
| BF939362.1 | 1 | 336 |
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | NM_001289164.1 | 12 | 2072 | NP_001276093.1 |
| NM_001289165.1 | 10 | 1789 | NP_001276094.1 | |
| NM_032603.3 | 14 | 2507 | NP_115992.1 | |
| XM_011533134.2 | 14 | 2467 | XP_011531436.1 | |
| XM_017005112.1 | 12 | 2111 | XP_016860601.1 | |
| AF282619.1 | 14 | 2480 | AAK51671.1 | |
| AF284815.1 | 14 | 2487 | AAK91134.1 | |
| AF311313.1 | 14 | 2478 | AAK63205.1 | |
| AK309478.1 | 12 | 2688 | ||
| BC033130.1 | 7 | 1013 | AAH33130.1 | |
| BC071865.1 | 12 | 2096 | AAH71865.1 | |
| DQ378059.1 | 10 | 1789 | ABD23013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map