Assay Details
Target Gene Details
Entrez Gene ID: | 57124 |
Gene Name: | CD248 molecule |
Gene Aliases: |
CD164L1, TEM1 |
Location: |
Chr.11:66314487-66317044 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD248 | NM_020404.2 | 1 | 1187 | NP_065137.1 |
| AF279142.1 | 1 | 1175 | AAG00867.1 | |
| AJ295846.1 | 1 | 1187 | CAC34381.1 | |
| AK027290.1 | 1 | 309 | BAB55018.1 | |
| BC051340.1 | 1 | 1174 | AAH51340.1 | |
| BC104484.1 | 1 | 1187 | AAI04485.1 | |
| BC105633.1 | 1 | 1187 | AAI05634.1 | |
| KU215671.1 | 1 | 1125 | ||
| KU257670.1 | 1 | 1119 |
More Information
Additional Information:
For this assay, SNP(s) [rs114284078,rs148943512] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map