Assay Details
Target Gene Details
Entrez Gene ID: | 5427 |
Gene Name: | DNA polymerase epsilon 2, accessory subunit |
Gene Aliases: |
DPE2 |
Location: |
Chr.14:49643552-49688417 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLE2 | NM_001197330.1 | NP_001184259.1 | ||
| NM_001197331.1 | NP_001184260.1 | |||
| NM_002692.3 | NP_002683.2 | |||
| XM_011536842.2 | XP_011535144.1 | |||
| XM_011536843.2 | XP_011535145.1 | |||
| XM_017021374.1 | XP_016876863.1 | |||
| XM_017021375.1 | XP_016876864.1 | |||
| AF025840.1 | AAC51920.1 | |||
| AF036899.1 | AAC39610.1 | |||
| AK293163.1 | ||||
| AK314963.1 | ||||
| BC112962.1 | ||||
| BC126218.1 | ||||
| BC126220.1 | ||||
| BX248064.1 | CAD62358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035708 | Chr.14:49632775 - 49749281 on Build GRCh38 | Gain |
|
| esv3634466 | Chr.14:49643688 - 49728973 on Build GRCh38 | Gain |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map