Assay Details
Target Gene Details
Entrez Gene ID: | 6196 |
Gene Name: | ribosomal protein S6 kinase A2 |
Gene Aliases: |
HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, p90RSK2, pp90RSK3 |
Location: |
Chr.6:166409364-166862551 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA2 | NM_001006932.2 | 22 | 5850 | NP_001006933.2 |
| NM_001318936.1 | 23 | 5901 | NP_001305865.1 | |
| NM_001318937.1 | 19 | 5558 | NP_001305866.1 | |
| NM_001318938.1 | 21 | 5437 | NP_001305867.1 | |
| NM_021135.5 | 21 | 5703 | NP_066958.2 | |
| XM_006715549.3 | 22 | 5732 | XP_006715612.1 | |
| AB073884.1 | 1 | 1699 | ||
| AB209116.1 | 23 | 5664 | BAD92353.1 | |
| AF140710.1 | 1 | 708 | ||
| BQ029058.1 | 1 | 169 | ||
| BU617697.1 | 1 | 129 |
More Information
Set Membership: |
|

Genomic Map