Assay Details
Target Gene Details
Entrez Gene ID: | 4802 |
Gene Name: | nuclear transcription factor Y subunit gamma |
Gene Aliases: |
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC |
Location: |
Chr.1:40691570-40771603 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYC | NM_001142587.1 | NP_001136059.1 | ||
| NM_001142588.1 | NP_001136060.1 | |||
| NM_001142589.1 | NP_001136061.1 | |||
| NM_014223.4 | NP_055038.2 | |||
| XM_006710661.1 | XP_006710724.1 | |||
| XM_017001365.1 | XP_016856854.1 | |||
| AB208975.1 | BAD92212.1 | |||
| AK300774.1 | ||||
| AK301215.1 | ||||
| AK301385.1 | ||||
| BC005003.1 | AAH05003.1 | |||
| D85425.1 | BAA12818.1 | |||
| D89986.1 | BAA14051.1 | |||
| DC391720.1 | ||||
| DC428152.1 | ||||
| U78774.1 | AAC51669.1 | |||
| Z70024.1 | CAA93846.1 | |||
| Z74792.1 | CAA99055.1 |
Target Gene Details
Entrez Gene ID: | 100130557 |
Gene Name: | NFYC antisense RNA 1 |
Gene Aliases: |
0808y08y |
Location: |
Chr.1:40689080-40692261 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYC-AS1 | NR_024567.1 | 1 | 381 | |
| AK094323.1 | 1 | 186 | BAC04330.1 | |
| DA926154.1 | 1 | 382 |
Target Gene Details
Entrez Gene ID: | 9783 |
Gene Name: | regulating synaptic membrane exocytosis 3 |
Gene Aliases: |
NIM3, RIM3 |
Location: |
Chr.1:40620680-40692104 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMS3 | XM_011542479.2 | XP_011540781.1 |
More Information
Set Membership: |
|

Genomic Map