Assay Details
Target Gene Details
Entrez Gene ID: | 2978 |
Gene Name: | guanylate cyclase activator 1A |
Gene Aliases: |
C6orf131, COD3, CORD14, GCAP, GCAP1, GUCA, GUCA1 |
Location: |
Chr.6:42155377-42180083 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCA1A | NM_000409.4 | 1 | 108 | NP_000400.2 |
| NM_001319061.1 | 1 | 108 | NP_001305990.1 | |
| NM_001319062.1 | 1 | 108 | NP_001305991.1 | |
| XM_011514537.2 | XP_011512839.1 | |||
| XM_011514539.1 | XP_011512841.1 | |||
| AA400590.1 | ||||
| AK125780.1 | ||||
| AK313865.1 | 1 | 79 | ||
| BC031663.1 | 1 | 49 | AAH31663.1 | |
| DB049592.1 | 1 | 79 | ||
| DB055332.1 | 1 | 108 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830649 | Chr.6:42047596 - 42231697 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map