Assay Details
Target Gene Details
Entrez Gene ID: | 5522 |
Gene Name: | protein phosphatase 2 regulatory subunit Bgamma |
Gene Aliases: |
B55-GAMMA, IMYPNO, IMYPNO1, PR52, PR55G |
Location: |
Chr.4:6320578-6563600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2C | NM_001206994.1 | NP_001193923.1 | ||
| NM_001206995.1 | NP_001193924.1 | |||
| NM_001206996.1 | NP_001193925.1 | |||
| NM_020416.3 | NP_065149.2 | |||
| NM_181876.2 | NP_870991.1 | |||
| XM_005247979.4 | XP_005248036.1 | |||
| XM_011513495.1 | XP_011511797.1 | |||
| XM_011513496.1 | XP_011511798.1 | |||
| AB209183.1 | BAD92420.1 | |||
| AF086924.1 | AAG39636.1 | |||
| AF145027.1 | AAP97279.1 | |||
| AK093115.1 | ||||
| AK122849.1 | ||||
| AK296474.1 | ||||
| AK316003.1 | ||||
| BC032954.2 | AAH32954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469711 | Chr.4:6210020 - 6384370 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map