Assay Details
Target Gene Details
Entrez Gene ID: | 9267 |
Gene Name: | cytohesin 1 |
Gene Aliases: |
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7 |
Location: |
Chr.17:78674047-78782342 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYTH1 | NM_001292018.1 | 14 | 2982 | NP_001278947.1 |
| NM_001292019.1 | 14 | 2548 | NP_001278948.1 | |
| NM_004762.3 | 14 | 2585 | NP_004753.1 | |
| NM_017456.3 | 13 | 2582 | NP_059430.2 | |
| XM_011525475.2 | 13 | 3325 | XP_011523777.1 | |
| XM_011525476.2 | 14 | 2500 | XP_011523778.1 | |
| XM_011525477.1 | 12 | 2363 | XP_011523779.1 | |
| XM_011525478.1 | 13 | 2904 | XP_011523780.1 | |
| XM_017025336.1 | 13 | 2652 | XP_016880825.1 | |
| XM_017025338.1 | 13 | 2968 | XP_016880827.1 | |
| AF054997.1 | 1 | 945 | ||
| AK091369.1 | 1 | 2084 | ||
| AK123894.1 | 1 | 3650 | ||
| AL832791.1 | 1 | 2763 | ||
| BC038385.1 | 14 | 2506 | AAH38385.1 | |
| BC050452.1 | 14 | 2495 | AAH50452.1 | |
| M85169.1 | 14 | 2535 | AAA36602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3285n100 | Chr.17:78595683 - 78678139 on Build GRCh38 | Loss |
|
| nsv482920 | Chr.17:78607335 - 78727244 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map