Assay Details
Target Gene Details
Entrez Gene ID: | 57704 |
Gene Name: | glucosylceramidase beta 2 |
Gene Aliases: |
AD035, NLGase, SPG46 |
Location: |
Chr.9:35736862-35749255 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
More Information
Set Membership: |
|

Genomic Map