Assay Details
Target Gene Details
Entrez Gene ID: | 26575 |
Gene Name: | regulator of G-protein signaling 17 |
Gene Aliases: |
RGS-17, RGSZ2, hRGS17 |
Location: |
Chr.6:153010891-153131284 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS17 | NM_012419.4 | 5 | 1091 | NP_036551.3 |
| AA902430.1 | ||||
| AF202257.3 | 5 | 1102 | AAF08978.3 | |
| BC013117.1 | 5 | 1086 | AAH13117.1 | |
| CK902282.1 | 1 | 382 |
More Information
Set Membership: |
|

Genomic Map