Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK1 | NM_001320923.1 | 24 | 3436 | NP_001307852.1 |
| NM_001321852.1 | 23 | 3439 | NP_001308781.1 | |
| NM_001321853.1 | 25 | 3698 | NP_001308782.1 | |
| NM_001321854.1 | 24 | 3614 | NP_001308783.1 | |
| NM_001321855.1 | 24 | 3617 | NP_001308784.1 | |
| NM_001321856.1 | 23 | 3352 | NP_001308785.1 | |
| NM_001321857.1 | 23 | 3530 | NP_001308786.1 | |
| NM_002227.3 | 23 | 3533 | NP_002218.2 | |
| XM_017001212.1 | 23 | 3377 | XP_016856701.1 | |
| AB209057.1 | 22 | 3289 | BAD92294.1 | |
| AB219242.1 | 21 | 3180 | BAE02826.1 | |
| AK314257.1 | 24 | 3437 | ||
| BC132729.1 | 22 | 3251 | ||
| BX647675.1 | 9 | 1137 | ||
| HQ628631.1 | 21 | 3213 | ||
| M64174.1 | 22 | 3258 | AAA36527.1 |
More Information
Set Membership: |
|

Genomic Map