Assay Details
Target Gene Details
Entrez Gene ID: | 80024 |
Gene Name: | solute carrier family 8 member B1 |
Gene Aliases: |
NCKX6, NCLX, SLC24A6 |
Location: |
Chr.12:113298766-113335142 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8B1 | NM_024959.2 | NP_079235.2 | ||
| XM_006719607.2 | XP_006719670.1 | |||
| XM_011538749.2 | XP_011537051.1 | |||
| XM_011538750.2 | XP_011537052.1 | |||
| XM_011538752.2 | XP_011537054.1 | |||
| AK025886.1 | BAB15271.1 | |||
| AK094074.1 | ||||
| AK095092.1 | ||||
| AK293216.1 | ||||
| AY601759.1 | AAT35807.1 | |||
| AY601760.1 | AAT35808.1 | |||
| BC098360.1 | AAH98360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map