Assay Details
Target Gene Details
Entrez Gene ID: | 2582 |
Gene Name: | UDP-galactose-4-epimerase |
Gene Aliases: |
SDR1E1 |
Location: |
Chr.1:23795599-23800804 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALE | NM_000403.3 | 4 | 423 | NP_000394.2 |
| NM_001008216.1 | 4 | 358 | NP_001008217.1 | |
| NM_001127621.1 | 3 | 399 | NP_001121093.1 | |
| AK057302.1 | 3 | 210 | ||
| AK096127.1 | 2 | 1737 | ||
| AK307354.1 | 4 | 308 | ||
| AK314397.1 | 3 | 371 | ||
| AL530475.3 | 4 | 358 | ||
| BC001273.1 | 3 | 279 | AAH01273.1 | |
| BC050685.2 | 4 | 282 | AAH50685.2 | |
| BQ437612.1 | 4 | 481 | ||
| DA043957.1 | 2 | 273 | ||
| DQ233668.1 | 2 | 189 | ABB04110.1 | |
| L41668.1 | 4 | 282 | AAB86498.1 |
More Information
Set Membership: |
|

Genomic Map