Assay Details
Target Gene Details
Entrez Gene ID: | 9457 |
Gene Name: | four and a half LIM domains 5 |
Gene Aliases: |
1700027G07Rik, ACT, FHL-5, dJ393D12.2 |
Location: |
Chr.6:96562525-96618626 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL5 | NM_001170807.2 | 1 | 75 | NP_001164278.1 |
| NM_020482.5 | 1 | 75 | NP_065228.4 | |
| AK058175.1 | 1 | 41 | ||
| AK223539.1 | BAD97259.1 | |||
| BC021723.2 | AAH21723.1 | |||
| BC029490.1 | AAH29490.1 | |||
| HY020942.1 | 1 | 76 |
More Information
Set Membership: |
|

Genomic Map