Assay Details
Target Gene Details
Entrez Gene ID: | 9088 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
MYT1, PPP1R126 |
Location: |
Chr.16:2972791-2980539 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKMYT1 | NM_001258450.1 | NP_001245379.1 | ||
| NM_001258451.1 | 3 | 566 | NP_001245380.1 | |
| NM_004203.4 | 3 | 702 | NP_004194.3 | |
| NM_182687.2 | 3 | 702 | NP_872629.1 | |
| XM_011522734.2 | 3 | 663 | XP_011521036.1 | |
| XM_011522735.2 | 3 | 489 | XP_011521037.1 | |
| XM_011522736.2 | 2 | 496 | XP_011521038.1 | |
| AK097642.1 | 3 | 451 | ||
| AK098452.1 | 3 | 554 | ||
| AK294379.1 | 2 | 463 | ||
| AK301926.1 | ||||
| AK303005.1 | 3 | 580 | ||
| AK309964.1 | 2 | 496 | ||
| BC121161.1 | 2 | 390 | ||
| BC121162.1 | 2 | 390 | ||
| BC130011.1 | 2 | 390 | ||
| U56816.1 | 2 | 406 | AAC50949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510673 | Chr.16:2919880 - 3043776 on Build GRCh38 | Deletion |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map