Assay Details
Target Gene Details
Entrez Gene ID: | 26103 |
Gene Name: | leucine rich repeat, Ig-like and transmembrane domains 1 |
Gene Aliases: |
FIGLER9, LRRC21, PAL |
Location: |
Chr.10:84231520-84241461 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIT1 | NM_015613.2 | 2 | 557 | NP_056428.1 |
| XM_011539623.2 | 1 | 813 | XP_011537925.1 | |
| AB031547.1 | 2 | 557 | BAA95681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain |
|
| nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map