Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGER4 | XM_017009659.1 | XP_016865148.1 |
Target Gene Details
Entrez Gene ID: | 23548 |
Gene Name: | tetratricopeptide repeat domain 33 |
Gene Aliases: |
OSRF |
Location: |
Chr.5:40711576-40756965 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC33 | NM_012382.2 | 5 | 3615 | NP_036514.1 |
| BE869916.1 | 1 | 377 |
More Information
Set Membership: |
|

Genomic Map