Assay Details
Target Gene Details
Entrez Gene ID: | 80332 |
Gene Name: | ADAM metallopeptidase domain 33 |
Gene Aliases: |
C20orf153, DJ964F7.1 |
Location: |
Chr.20:3667973-3682131 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575355 | Chr.20:3667953 - 3752840 on Build GRCh38 | Gain |
|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map