Assay Details
Target Gene Details
Entrez Gene ID: | 8624 |
Gene Name: | proteasome assembly chaperone 1 |
Gene Aliases: |
C21LRP, DSCR2, LRPC21, PAC-1, PAC1 |
Location: |
Chr.21:39175446-39183514 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG1 | NM_001261824.1 | 5 | 715 | NP_001248753.1 |
| NM_001320795.1 | 5 | 918 | NP_001307724.1 | |
| NM_003720.3 | 5 | 730 | NP_003711.1 | |
| NM_203433.2 | 4 | 667 | NP_982257.1 | |
| NR_049728.1 | 4 | 623 | ||
| XM_017028488.1 | 4 | 581 | XP_016883977.1 | |
| AB451468.1 | 4 | 538 | ||
| AF417108.1 | 4 | 494 | ||
| AJ006291.1 | 5 | 664 | CAA06957.1 | |
| AK001455.1 | 5 | 692 | ||
| AK308614.1 | 4 | 601 | ||
| AK315406.1 | 5 | 632 | ||
| AW594615.1 | 3 | 411 | ||
| AY463963.1 | 4 | 538 | AAR25628.1 | |
| BC003619.2 | 5 | 647 | AAH03619.1 | |
| BC010424.1 | 5 | 639 | AAH10424.1 | |
| BC011755.2 | 5 | 647 | AAH11755.1 | |
| BC012809.2 | 5 | 647 | AAH12809.1 | |
| BE779375.1 | ||||
| BM013494.1 | 5 | 600 | ||
| BM460414.1 | 5 | 623 | ||
| BM462074.1 | 5 | 647 | ||
| BQ008489.1 | 3 | 415 | ||
| BU543452.1 | ||||
| CB160221.1 | ||||
| CR541821.1 | 5 | 601 | CAG46620.1 | |
| CR541852.1 | 5 | 601 | CAG46650.1 |
More Information
Set Membership: |
|

Genomic Map