Assay Details
Target Gene Details
Entrez Gene ID: | 103 |
Gene Name: | adenosine deaminase, RNA specific |
Gene Aliases: |
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136 |
Location: |
Chr.1:154582057-154628016 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAR | NM_001025107.2 | 7 | 2461 | NP_001020278.1 |
| NM_001111.4 | 7 | 2592 | NP_001102.2 | |
| NM_001193495.1 | 7 | 2630 | NP_001180424.1 | |
| NM_015840.3 | 7 | 2592 | NP_056655.2 | |
| NM_015841.3 | 7 | 2535 | NP_056656.2 | |
| XM_006711109.2 | 7 | 2398 | XP_006711172.1 | |
| XM_006711111.3 | 8 | 2520 | XP_006711174.1 | |
| XM_006711112.2 | 8 | 2222 | XP_006711175.1 | |
| XM_006711113.2 | 8 | 2296 | XP_006711176.1 | |
| XM_011509060.1 | 7 | 2494 | XP_011507362.1 | |
| XM_011509061.2 | 7 | 2461 | XP_011507363.2 | |
| XM_011509062.1 | 7 | 2421 | XP_011507364.1 | |
| XM_017000037.1 | 8 | 2389 | XP_016855526.1 | |
| AB209891.1 | 7 | 2405 | BAD93128.1 | |
| AK304153.1 | 7 | 2546 | ||
| BC038227.1 | 7 | 2519 | AAH38227.1 | |
| BU430287.1 | ||||
| BX538232.1 | 7 | 2445 | CAD98075.1 | |
| BX640741.1 | 7 | 2443 | CAE45853.1 | |
| KJ544856.1 | 6 | 2350 | ||
| U10439.1 | 7 | 2504 | AAB06697.1 | |
| U18121.1 | 7 | 2397 | AAC13782.1 | |
| X79448.1 | 7 | 2630 | CAA55967.1 | |
| X79449.1 | 7 | 2552 | CAA55968.1 | |
| X98559.1 | 6 | 2335 | CAA67170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map