Assay Details
Target Gene Details
Entrez Gene ID: | 4129 |
Gene Name: | monoamine oxidase B |
Gene Aliases: |
- |
Location: |
Chr.X:43766610-43882475 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOB | NM_000898.4 | 15 | 1672 | NP_000889.3 |
| XM_005272608.3 | 17 | 1772 | XP_005272665.1 | |
| XM_017029523.1 | 17 | 1822 | XP_016885012.1 | |
| XM_017029524.1 | 16 | 1632 | XP_016885013.1 | |
| AK294306.1 | 16 | 1590 | ||
| AK298942.1 | 14 | 1511 | ||
| AK312679.1 | 15 | 1647 | ||
| BC022494.1 | 15 | 1618 | AAH22494.1 | |
| CA975968.1 | 2 | 133 | ||
| M69177.1 | 15 | 1573 | AAA59550.1 | |
| S62734.1 | 15 | 1496 | AAB27229.1 |
More Information
Set Membership: |
|

Genomic Map