Assay Details
Target Gene Details
Entrez Gene ID: | 10289 |
Gene Name: | eukaryotic translation initiation factor 1B |
Gene Aliases: |
GC20 |
Location: |
Chr.3:40309682-40312424 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1B | NM_005875.2 | NP_005866.1 | ||
| AF077052.1 | AAD27785.1 | |||
| AF263452.1 | AAF79182.1 | |||
| AK223448.1 | BAD97168.1 | |||
| BC006996.1 | AAH06996.1 | |||
| BF667107.1 | ||||
| CA335728.1 |
Target Gene Details
Entrez Gene ID: | 440952 |
Gene Name: | EIF1B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:40173145-40309698 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1B-AS1 | NR_033965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834667 | Chr.3:40149053 - 40317061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113060212] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map