Assay Details
Target Gene Details
Entrez Gene ID: | 1495 |
Gene Name: | catenin alpha 1 |
Gene Aliases: |
CAP102, MDPT2 |
Location: |
Chr.5:138753386-138935034 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNA1 | NM_001290307.2 | NP_001277236.1 | ||
| NM_001290309.2 | NP_001277238.1 | |||
| NM_001290310.2 | NP_001277239.1 | |||
| NM_001323982.1 | NP_001310911.1 | |||
| NM_001323983.1 | NP_001310912.1 | |||
| NM_001323984.1 | NP_001310913.1 | |||
| NM_001323985.1 | NP_001310914.1 | |||
| NM_001323986.1 | NP_001310915.1 | |||
| NM_001903.4 | NP_001894.2 | |||
| AK094151.1 | ||||
| AK296708.1 | ||||
| AK300439.1 | ||||
| AK304267.1 | ||||
| BC000385.2 | AAH00385.1 | |||
| D13866.1 | BAA02979.1 | |||
| D14705.1 | BAA03530.1 | |||
| L23805.1 | AAA86430.1 | |||
| U03100.1 | AAA18949.1 |
Target Gene Details
Entrez Gene ID: | 26045 |
Gene Name: | leucine rich repeat transmembrane neuronal 2 |
Gene Aliases: |
- |
Location: |
Chr.5:138869390-138875368 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRTM2 | NM_015564.2 | 2 | 5123 | NP_056379.1 |
| AB007876.1 | 2 | 5075 |
More Information
Set Membership: |
|

Genomic Map