Assay Details
Target Gene Details
Entrez Gene ID: | 9136 |
Gene Name: | ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast) |
Gene Aliases: |
RNU3IP2, U3-55K |
Location: |
Chr.3:51933426-51941941 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP9 | NM_004704.4 | NP_004695.1 | ||
| AJ001340.1 | CAA04687.1 | |||
| AK313696.1 | ||||
| BC001113.1 | AAH01113.1 | |||
| BC009879.2 | AAH09879.1 | |||
| BC010048.1 | AAH10048.1 | |||
| BC023662.2 | AAH23662.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
| nsv590280 | Chr.3:51933423 - 51944311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map