Assay Details
Target Gene Details
Entrez Gene ID: | 8829 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
BDCA4, CD304, NP1, NRP, VEGF165R |
Location: |
Chr.10:33177491-33334905 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRP1 | NM_001244972.1 | 17 | 3174 | NP_001231901.1 |
| NM_001244973.1 | 17 | 3171 | NP_001231902.1 | |
| NM_003873.5 | 17 | 3192 | NP_003864.4 | |
| NR_045259.1 | 16 | 2989 | ||
| XM_006717521.1 | 17 | 2969 | XP_006717584.1 | |
| XM_006717522.1 | 16 | 2918 | XP_006717585.1 | |
| XM_006717523.2 | 16 | 2915 | XP_006717586.1 | |
| XM_006717524.1 | 16 | 2864 | XP_006717587.1 | |
| XM_006717525.1 | 16 | 2861 | XP_006717588.1 | |
| XM_017016865.1 | 16 | 2894 | XP_016872354.1 | |
| XM_017016866.1 | 16 | 2786 | XP_016872355.1 | |
| AB209641.1 | 17 | 2820 | BAD92878.1 | |
| AF016050.1 | 17 | 2669 | AAC12921.1 | |
| AF018956.1 | 17 | 2669 | AAC51759.1 | |
| AK292822.1 | 17 | 2840 | ||
| AL833608.1 | 16 | 2720 | CAH10397.1 | |
| AY249243.1 | 16 | 2618 | AAP80144.1 | |
| BX510902.1 | 17 | 2856 | CAD91133.1 | |
| BX648250.1 | 17 | 3140 | ||
| CR749333.1 | 17 | 3193 | CAH18187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map